Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM87B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525209

Catalog No. PIPA525209


Add to cart

Specifications

Specifications

TMEM87B
Polyclonal
PBS with 0.09% sodium azide
Q96K49, Q8BKU8
TMEM87B
KLH conjugated synthetic peptide between 484-513 amino acids from the C-terminal region of human TMEM87B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
Unconjugated
TMEM87B
Liquid
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
72477, 84910
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit