Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-TMEM97, Alexa Fluor 488, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP130437AF488
Description
TMEM97 Polyclonal antibody specifically detects TMEM97 in Mouse samples. It is validated for Western Blot, ICC/IF (Negative).Specifications
TMEM97 | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Affinity Purified | |
TMEM97 | |
Synthetic peptide made to an internal portion of the mouse TMEM97 protein (within residues 20-100. [Swiss-Prot# Q8VD00] | |
24 kDa | |
0.1 ml | |
Cholesterol Metabolism, Cytokine Research, Ovarian Carcinoma Cell Markers | |
Primary | |
27346 | |
Mouse |
Immunocytochemistry, Western Blot | |
Western Blot, ICC/IF (Negative) | |
Q5BJF2 | |
MAC30Protein MAC30, transmembrane protein 97 | |
Rabbit | |
IgG | |
Immunogen affinity purified | |
RUO | |
Store at 4C in the dark. | |
Polyclonal | |
Mouse. Immunogen sequence has 93% homology with rat. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only