Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TMEM97, Alexa Fluor 647, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130436AF647

Catalog No. NBP130436Y


Add to cart

Description

Description

TMEM97 Polyclonal antibody specifically detects TMEM97 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

TMEM97
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
TMEM97
Synthetic peptide made to an internal portion of the human TMEM97 protein (within residues 105-155. [Swiss-Prot: Q5BJF2]
24 kDa
0.1 ml
Cholesterol Metabolism, Cytokine Research, Ovarian Carcinoma Cell Markers
Primary
27346
Human
Immunocytochemistry, Immunofluorescence, Western Blot
Western Blot, Immunocytochemistry/Immunofluorescence
Q5BJF2
MAC30Protein MAC30, transmembrane protein 97
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human. Predicted to react with monkey based on 100% sequence homology.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only