Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM97 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | TMEM97 |
---|---|
Concentration | 1 mg/ml |
Dilution | Western Blot 2 ug/ml, ICC/IF (Negative) |
Applications | Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP130437SS
|
Novus Biologicals
NBP130437SS |
0.025 mL |
Each for $204.00
|
|
|||||
NBP130437
|
Novus Biologicals
NBP130437 |
0.1 mL |
Each for $482.50
|
|
|||||
Description
TMEM97 Polyclonal specifically detects TMEM97 in Mouse samples. It is validated for Western Blot, ICC/IF (Negative).Specifications
TMEM97 | |
Western Blot 2 ug/ml, ICC/IF (Negative) | |
Polyclonal | |
Rabbit | |
Cholesterol Metabolism, Cytokine Research, Ovarian Carcinoma Cell Markers | |
PBS with 0.02% Sodium Azide | |
MAC30Protein MAC30, transmembrane protein 97 | |
TMEM97 | |
IgG | |
Affinity Purified | |
Mouse. Immunogen sequence has 93% homology with rat. |
1 mg/ml | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
RUO | |
Mouse | |
Q5BJF2, Q8VD00 | |
27346 | |
Synthetic peptide made to an internal portion of the mouse TMEM97 protein (within residues 20-100. [Swiss-Prot# Q8VD00] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
24 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title