Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMEM97 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523004

Catalog No. PIPA523004

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The target sequence has 93% sequence homology with rat. This antibody is not recommended for ICC/IF applications.

TMEM97, or MAC30, is a conserved transmembrane protein that helps regulate cellular cholesterol levels through colocalization with NPC1. TMEM97 has also been linked to several cancers including breast, pancreatic, colorectal and ovarian cancers.
Specifications

Specifications

TMEM97
Polyclonal
Unconjugated
Tmem97
Liquid
TMEM97
Synthetic peptide made to an internal portion of the mouse protein (within residues 20-100.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Western Blot
1.1 mg/mL
PBS with 30% glycerol and 0.1% sodium azide
Q8VD00
MAC30
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
69071
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit