Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMLHE Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524235

Catalog No. PIPA524235

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and mouse based on sequence homology.

This gene encodes the protein trimethyllysine dioxygenase which is the first enzyme in the carnitine biosynthesis pathway. Carnitine play an essential role in the transport of activated fatty acids across the inner mitochondrial membrane. The encoded protein converts trimethyllysine into hydroxytrimethyllysine. A pseudogene of this gene is found on chromosome X. Alternate splicing results in multiple transcript variants.
Specifications

Specifications

TMLHE
Polyclonal
PBS with 0.09% sodium azide
Q9NVH6
BBOX2, TMLD, TMLH, XAP130, TML hydroxylase, TML-alpha-ketoglutarate dioxygenase, butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetaine hydroxylase) 2, epsilon-trimethyllysine 2-oxoglutarate dioxygenase, epsilon-trimethyllysine hydroxylase, trimethyllysine dioxygenase, m
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55217
Western Blot
Unconjugated
TMLHE
Liquid
TMLHE
KLH conjugated synthetic peptide between 120-149 amino acids from the N-terminal region of human TMLHE
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit