Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TMOD1, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127777

Catalog No. PIPA127777

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Blocks the elongation and depolymerization of the actin filaments at the pointed end. The Tmod/TM complex contributes to the formation of the short actin protofilament. which in turn defines the geometry of the membrane skeleton. May play an important rol

PA1-27777 detects TMOD1 from human, mouse, and rat samples.PA1-27777 may be successfully used in ELISA and Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Tropomodulin-1.
Specifications

Specifications

TMOD1
Polyclonal
Unconjugated
TMOD1
Purified
TMOD1
Synthetic peptide corresponding to human Tropomodulin-1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
ELISA, Western Blot
1mg/mL
PBS with no preservative
P28289, P49813, P70567
D9S57E, ETMOD, TMOD
Chicken
IgY
50μg
Store at -20°C
Primary
7111
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit