Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMOD4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA545817
Description
Peptide sequence: CNTEGISSVV QPDKYKPVPD EPPNPTNIEE ILKRVRSNDK ELEEVNLNNI Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 85%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 93%; Rat: 100%
TMOD4 Polyclonal antibody specifically detects TMOD4 in Human samples. It is validated for Western Blot.Specifications
TMOD4 | |
Polyclonal | |
Unconjugated | |
TMOD4 | |
actin-capping protein; MTMOD; skeletal muscle tropomodulin; skeletal tropomodulin; SK-TMOD; TMOD4; tropomodulin 4; tropomodulin 4 (muscle); tropomodulin-4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
29765 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NZQ9 | |
TMOD4 | |
Synthetic peptide directed towards the middle region of human TMOD4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction