Learn More
MilliporeSigma™ anti-TMPRSS2 Clone: 2F4, Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma™ ST1676100UG
Catalog No. ST1676100UG
Add to cart
Description
Description
Specifically detects TMPRSS2 Clone: 2F4 in Human samples, and it is validated for ELISA, Immuno Blotting
This gene encodes a protein that belongs to the serine protease family. The encoded protein contains a type II transmembrane domain, a receptor class A domain, a scavenger receptor cysteine-rich domain and a protease domain. Serine proteases are known to be involved in many physiological and pathological processes. This gene was demonstrated to be up-regulated by androgenic hormones in prostate cancer cells and down-regulated in androgen-independent prostate cancer tissue. The protease domain of this protein is thought to be cleaved and secreted into cell media after autocleavage. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
TMPRSS2
2F4
In PBS, pH 7.2.
Murine
IgG2a
RUO
Primary
Recognizes the TMPRSS2 protein.
ELISA, Immuno Blotting, Immunoblot
Unconjugated
Purified
A recombinant polypeptide corresponding to amino acids of 383-493 human TMPRSS2, expressed as a GST fusion protein
100μg
−20°C
Monoclonal
Human