Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYCARD Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141405

Catalog No. 01-675-534


Add to cart

Description

Description

Suggested positive control: A549, antigen standard for PYCARD (transient overexpression lysate).

TMS1 (target of methylation-induced silencing) is a recently identified CpG island-associated gene that becomes hypermethylated and silenced in cells overexpressing DNA cytosine-5-methyltransferase-1 (1). TMS1 is aberrantly methylated and silenced in human breast cancer cells. Forty percent (11 of 27) of primary breast tumors exhibited aberrant methylation of TMS1. Ectopic expression of TMS1 induced apoptosis in 293 cells and inhibited the survival of human breast cancer cells. The methylation-mediated silencing of TMS1 may confer a survival advantage by allowing cells to escape from apoptosis, supporting a new role for aberrant methylation in breast tumorigenesis. TMS1 encodes a 22-kDa predicted protein containing a COOH-terminal caspase recruitment domain (CARD). However, it is structurally unrelated to other known CARD adaptor and regulatory proteins. Ectopic expression of TMS1 alone was able to trigger apoptosis in 293 cells, and cell death correlated with relocalization of TMS1 from the cytoplasm to perinuclear, ball-like structures. Several lines of evidence support the idea that redistribution of TMS1 is an intermediate step in a TMS1-triggered apoptotic pathway (2).
Specifications

Specifications

PYCARD
Polyclonal
Unconjugated
PYCARD
Liquid
PYCARD
Synthetic peptides corresponding to residues 31-46 and 94-110 of human TMS1.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
antibody with 0.2% gelatin and 0.02% sodium azide
Q9ULZ3
ASC, CARD5, MGC10332, TMS, TMS-1, TMS1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
29108
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit