Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF, Rat anti-Mouse, Clone: MP6-XT22, BV711, BD

Manufacturer:  BD Biosciences 563944

Catalog No. BDB563944


Add to cart

Description

Description

Host Species: Rat
Clone: MP6-XT22
Isotype: IgG1
Species Reactivity: Ms
Immunogen: Recombinant Mouse TNF

Flow Cytometry (Intracellular Staining)

Specifications

Specifications

TNF
0.2mg/mL
Rat
50μg
Primary
Murine
MP6-XT22
Affinity Purified
Recombinant Mouse TNF
RUO
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit