Learn More
TNFR2, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA121148
Catalog No. PA121148
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Tumor Necrosis Factor (TNF) is a cytokine whose function is mediated through two distinct cell surface receptors (TNFR 1 and TNFR 2) that are included in the TNF receptor superfamily along with FAS antigen and CD40. Tumor necrosis factor receptor 1 and 2 (TNFR 1 and TNFR 2) are 55 and 75kDa members of a family cell surface molecules including nerve growth factor receptor, Fas/Apo1, CD30, Ox40, and 4-1BB, which are characterized by cysteine rich motifs in the extracellular domain.
PA1-21148 detects TNF receptor II in human, mouse and rat samples.PA1-21148 has been successfully used in immunohistochemistry (paraffin), Immunoprecipitation and Western blot procedures.The PA1-21148 immunogen is a synthetic peptide to the C-terminal region of mouse TNF receptor II protein.
Specifications
Specifications
TNFR2
Polyclonal
Unconjugated
TNFRSF1B
Liquid
TNFRSF1B
Synthetic peptide to the C-terminal region of mouse TNF receptor II protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.2mg/mL
PBS with 1% BSA and 0.09% sodium azide; pH 7.4
P20333, P25119, Q80WY6
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR
Rabbit
IgG
500μL
4° C, do not freeze
Primary
156767, 21938, 7133