Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TNF RI/TNFRSF1A, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP145906

Catalog No. NBP145906

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TNF RI/TNFRSF1A
0.2 mg/mL
Western Blot, Immunohistochemistry
P25118
CD120a, CD120a antigen, FPF, p55, p55-R, p60, TNFARMGC19588, TNF-R, TNF-R1, TNFR1TBP1, TNFR55, TNF-R55, TNFR60, TNF-RI, TNF-R-I, TNFR-I, tumor necrosis factor binding protein 1, Tumor necrosis factor receptor 1, tumor necrosis factor receptor 1A isoform beta, tumor necrosis factor receptor superfamily member 1A, tumor necrosis factor receptor superfamily, member 1A, tumor necrosis factor receptor type 1, Tumor necrosis factor receptor type I, tumor necrosis factor-alpha receptor
Rabbit
IgG
0.1 mg
Apoptosis, Cancer, Cytokine Research
Primary
7132
Bovine, Canine, Hamster, Human, Monkey, Mouse, Porcine, Primate, Rabbit, Rat, Sheep
Immunohistochemistry, Western Blot
Unconjugated
PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Affinity Purified
TNFRSF1A
Synthetic peptide surrounding amino acid 32 of human TNF-RI.
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
The antibody detects TNF-RI of human, mouse, rat, and bovine origins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only