Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF RII/TNFRSF1B Mouse anti-Human, Clone: 22221, R&D Systems™

Mouse Monoclonal Antibody

$125.00 - $839.00

Specifications

Antigen TNF RII/TNFRSF1B
Immunogen E. coli-derived recombinant human TNF RII Leu23-Asp257 Accession # P20333
Clone 22221
Applications Flow Cytometry, Neutralization, Immunocytochemistry
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB226SP
View Documents
R&D Systems
MAB226SP
25 ug
Each for $125.00
Add to cart
 
MAB226100
View Documents
R&D Systems
MAB226100
100 ug
Each for $387.50
Add to cart
 
MAB226500
View Documents
R&D Systems
MAB226500
500 ug
Each for $839.00
Add to cart
 
Description

Description

TNF RII/TNFRSF1B Monoclonal antibody MAB226 Specifically detects TNF RII/TNFRSF1B in Human samples. It is validated for Immunocytochemistry, Neutralization
Specifications

Specifications

TNF RII/TNFRSF1B
22221
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG2a
7133
RUO
E. coli-derived recombinant human TNF RII Leu23-Asp257 Accession # P20333
Flow Cytometry, Neutralization, Immunocytochemistry
Purified
P20333
Monoclonal
Primary
Detects human TNF RII/TNFRSF1B in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhFas, rhGITR, rhHVEM, rhLTR beta , rhNGF R, rhOPG, rhRANK, rhTNF RI, or recombinant mouse TNF RII is observed. Neutralizes the biological effect of rhTNF RII but not rhTNF RI.
Protein A or G purified from ascites
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit