Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFAIP1, Polyclonal, Invitrogen™

Catalog No. PIPA127724
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127724 50 μg
1 options

Catalog No. PIPA127724

Supplier: Thermo Scientific PA127724

Chicken Polyclonal Antibody

This protein was identified as a gene whose expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. Studies of a similar gene in mouse suggest that the expression of this gene is developmentally regulated in

PA1-27724 detects TNFAIP1 from human, mouse, and rat samples.PA1-27724 may be successfully used in ELISA and Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Tumor necrosis factor. alpha-induced protein 1. Endothelial.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFAIP1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene TNFAIP1
Gene Accession No. Q13829, O70479, Q7TNY1
Gene Alias B12, B61, EDP1, MGC2317
Gene Symbols TNFAIP1
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Tumor necrosis factor. alpha-induced protein 1. Endothelial.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7126
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.