Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TNFAIP2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133480

Catalog No. NBP133480


Add to cart

Description

Description

TNFAIP2 Polyclonal antibody specifically detects TNFAIP2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TNFAIP2
1.02mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1-100-1:1000
Affinity Purified
TNFAIP2
Recombinant protein encompassing a sequence within the center region of human TNFalpha-IP 2. The exact sequence is proprietary.
73 kDa
0.1 ml
Angiogenesis, Breast Cancer, Cancer
Primary
7127
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
B94, EXOC3L3, exocyst complex component 3-like 3, Primary response gene B94 protein, TNF alpha-induced protein 2, tumor necrosis factor alpha-induced protein 2, tumor necrosis factor, alpha-induced protein 2
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only