Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNFAIP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520728
Description
A suggested positive control is 293 cell lysate. PA5-20728 can be used with blocking peptide PEP-0842.
TNFAIP3 Polyclonal antibody specifically detects TNFAIP3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
TNFAIP3 | |
Polyclonal | |
Unconjugated | |
TNFAIP3 | |
A20; A20p37; A20p50; AISBL; OTU domain-containing protein 7C; OTUD7C; Putative DNA-binding protein A20; RP11-356I2.3; TNF alpha induced protein 3; TNF alpha-induced protein 3; TNFA1P2; TNFAIP3; TNFAIP3 (A20); Tnfip3; Tumor necrosis factor alpha-induced protein 3; tumor necrosis factor induced protein 3; tumor necrosis factor inducible protein A20; tumor necrosis factor, alpha induced protein 3; tumor necrosis factor, alpha-induced protein 3; zinc finger protein A20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21929, 683206, 7128 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P21580, Q60769 | |
TNFAIP3 | |
A 14 amino acid peptide near the carboxy terminus of human TNFAIP3. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction