Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNFAIP8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524449

Catalog No. PIPA524449

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Acts as a negative mediator of apoptosis and may play a role in tumor progression. Suppresses the TNF-mediated apoptosis by inhibiting caspase-8 activity but not the processing of procaspase-8, subsequently resulting in inhibition of BID cleavage and caspase-3 activation.
Specifications

Specifications

TNFAIP8
Polyclonal
PBS with 0.09% sodium azide
O95379
GG2-1, MDC-3.13, SCC-S2, SCCS2, NDED, NF-kappa-B-inducible DED-containing protein, TNF alpha-induced protein 8, TNF-induced protein GG2-1, head and neck tumor and metastasis-related protein, tumor necrosis factor alpha-induced protein 8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25816
Flow Cytometry, Western Blot
Unconjugated
TNFAIP8
Liquid
TNFAIP8
KLH conjugated synthetic peptide between 50-78 amino acids from the N-terminal region of human TNFAIP8
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit