Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNIP1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524503

Catalog No. PIPA524503


Add to cart

Description

Description

This antibody is predicted to react with mouse based on sequence homology.

Interacts with zinc finger protein A20/TNFAIP3 and inhibits TNF-induced NF-kappa-B-dependent gene expression by interfering with an RIP-or TRAF2-mediated transactivation signal. Increases cell surface CD4(T4) antigen expression. Interacts with HIV-1 matrix protein and is packaged into virions and overexpression can inhibit viral replication. May regulate matrix nuclear localization, both nuclear import of PIC (Preintegration complex) and export of GAG polyprotein and viral genomic RNA during virion production.
Specifications

Specifications

TNIP1
Polyclonal
PBS with 0.09% sodium azide
Q15025
ABIN-1, NAF1, VAN, nip40-1, HIV-1 Nef-interacting protein, Nef-associated factor 1 SNP, TNFAIP3-interacting protein 1, virion-associated nuclear shuttling protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10318
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TNIP1
Liquid
TNIP1
KLH conjugated synthetic peptide between 454-482 amino acids from the C-terminal region of human TNIP1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit