Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tankyrase 2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525233

Catalog No. PIPA525233


Add to cart

Description

Description

Tnks 2 may regulate vesicle trafficking and modulate the subcellular distribution of SLC2A4/GLUT4-vesicles. Tnks 2 has PARP activity and can modify TRF1, and thereby contribute to the regulation of telomere length.
Specifications

Specifications

Tankyrase 2
Polyclonal
PBS with 0.09% sodium azide
Q9H2K2, Q3UES3
PARP-5b, PARP-5c, PARP5B, PARP5C, TANK2, TNKL, pART6, TNKS-2, TRF1-interacting ankyrin-related ADP-ribose polymerase 2, poly [ADP-ribose] polymerase 5B, tankyrase 2, tankyrase II, tankyrase-2, tankyrase-like protein, tankyrase-related protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
74493, 80351
Flow Cytometry, Western Blot
Unconjugated
TNKS2
Liquid
TNKS2
KLH conjugated synthetic peptide between 1126-1156 amino acids from the C-terminal region of human Tnks 2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit