Learn More
troponin I type 2 (skeletal, fast), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human TNNI2 protein.
Supplier: Abnova Corporation H00007136D01P
Description
This gene encodes a fast-twitch skeletal muscle protein, a member of the troponin I gene family, and a component of the troponin complex including troponin T, troponin C and troponin I subunits. The troponin complex, along with tropomyosin, is responsible for the calcium-dependent regulation of striated muscle contraction. Mouse studies show that this component is also present in vascular smooth muscle and may play a role in regulation of smooth muscle function. In addition to muscle tissues, this protein is found in corneal epithelium, cartilage where it is an inhibitor of angiogenesis to inhibit tumor growth and metastasis, and mammary gland where it functions as a co-activator of estrogen receptor-related receptor alpha. This protein also suppresses tumor growth in human ovarian carcinoma. Mutations in this gene cause myopathy and distal arthrogryposis type 2B. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq
Sequence: MGDEEKRNRAITARRQHLKSVMLQIAATELEKEESRREAEKQNYLAEHCPPLHIPGSMSEVQELCKQLHAKIDAAEEEKYDMEVRVQKTSKELEDMNQKLFDLRGKFKRPPLRRVRMSADAMLKALLGSKHKVCMDLRANLKQVKKEDTEKERDLRDVGDWRKNIEEKSGMEGRKKMFESESSpecifications
troponin I type 2 (skeletal, fast) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human TNNI2 protein. | |
TNNI2 | |
AMCD2B/DA2B/FSSV | |
Rabbit | |
Affinity Purified | |
RUO | |
7136 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_003282 | |
TNNI2 | |
TNNI2 (NP_003273.1, 1 a.a. ∼ 182 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.