Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TNNI3K Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP132837
Description
TNNI3K Polyclonal specifically detects TNNI3K in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
TNNI3K | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
TNNI3K | |
Recombinant protein encompassing a sequence within the center region of human TNNI3K. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1.0mg/mL | |
Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
CARKCardiac ankyrin repeat kinase, EC 2.7.11.1, MGC142099, MGC33828, serine/threonine-protein kinase TNNI3K, TNNI3 interacting kinase, TNNI3-interacting kinase | |
Rabbit | |
93 kDa | |
100 μL | |
Protein Kinase | |
51086 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction