Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

troponin T type 3 (skeletal, fast), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human TNNT3 protein.

Manufacturer:  Abnova Corporation H00007140B01

Catalog No. 89-006-377


Add to cart

Description

Description

Sequence: MSDEEVEQVEEQYEEEEEAQEEEEVQEEEKPRPKLTAPKIPEGEKVDFDDIQKKRQNKDLMELQALIDSHFEARKKEEEELVALKERIEKRRAERAEQQRIRAEKERERQNRLAEEKARREEEDAKRRAEDDLKKKKALSSMGANYSSYLAKADQKRGKKQTAREMKKKILAERRKPLNIDHLGEDKLRDKAKELWETLHQLEIDKFEFGEKLKRQKYDITTLRSRIDQAQKHSKKAGTPAKGKVGGRWK
Specifications

Specifications

troponin T type 3 (skeletal, fast)
Unconjugated
No additive
NM_006757
AMCD2B|DA2B|DKFZp779M2348|FSSV
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
7140
Western Blot
Mouse polyclonal antibody raised against a full-length human TNNT3 protein.
TNNT3
Serum
TNNT3
TNNT3 (NP_006748, 1 a.a. ~ 250 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit