Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tensin 1, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human TNS1 protein.

Manufacturer:  Abnova Corporation H00007145B01P

Catalog No. 89-006-378


Add to cart

Description

Description

The protein encoded by this gene localizes to focal adhesions, regions of the plasma membrane where the cell attaches to the extracellular matrix. This protein crosslinks actin filaments and contains a Src homology 2 (SH2) domain, which is often found in molecules involved in signal transduction. This protein is a substrate of calpain II. A second transcript from this gene has been described, but its full length nature has not been determined. [provided by RefSeq

Sequence: MSVSRTMEDSCELDLVYVTERIIAVSFPSTANEENFRSNLREVAQMLKSKHGGNYLLFNLSERRPDITKLHAKVLEFGWPDLHTPALEKICSICKAMDTWLNADPHNVVVLHNKGNRGRIGVVIAAYMHYSNISASADQALDRFAMKRFYEDKIVPIGQPSQRRYVHYFSGLLSGSIKMNNKPLFLHHVIMHGIPNFESKGGCRPFLRIYQAMQPVYTSGIYNIPGDSQTSVCITIEPGLLLKGDILLKCYHKKFRSPARDVIFRVQFHTCAIHDLGVVFGKEDLDDAFKDDRFPEYGKVEFVFSYGPEKIQGMEHLENGPSVSVDYNTSDPLIRWDSYDNFSGHRDDGMEDGNKQNTNSQSIGSISGGLEDQYTWPDTHWPSQS
Specifications

Specifications

tensin 1
Unconjugated
In 1X PBS, pH 7.4
BC051304
DKFZp586K0617|MGC88584|MST091|MST122|MST127|MSTP122|MSTP127|MXRA6|TNS
Mouse
Protein A column
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
7145
Western Blot
Mouse polyclonal antibody raised against a full-length human TNS1 protein.
TNS1
Affinity Purified
TNS1
TNS1 (AAH51304, 1 a.a. ~ 385 a.a) full-length human protein.
50 ug
Signal Transduction
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit