Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOB1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112687

Catalog No. 01-674-831


Add to cart

Description

Description

A suggested positive control for this product is HeLa cell lysate.

TOB does not contain either SH2 or SH3 domains but is homologous with the N terminus of the anti-proliferative BTG-1 protein. Like BTG-1, TOB when exogenously expressed, is able to suppress the growth of NIH3T3 cells. This growth suppression is hindered by the presence of kinase-active p185erbB2.
Specifications

Specifications

TOB1
Polyclonal
PBS with 0.08% sodium azide
P50616
APRO6, MGC104792, MGC34446, PIG49, TOB, TROB, TROB1
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10140
Western Blot
Unconjugated
TOB1
Liquid
TOB1
Synthetic peptide corresponding to the TOB.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit