Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOB2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513265

Catalog No. PIPA513265

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

TOB2 belongs to the TOB /BTG1 family of antiproliferative proteins, which are involved in the regulation of cell cycle progression.
Specifications

Specifications

TOB2
Polyclonal
PBS with 0.09% sodium azide
Q14106
KIAA1663; TOB4; TROB2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10766
Immunohistochemistry, Western Blot
Unconjugated
TOB2
Liquid
TOB2
KLH conjugated synthetic peptide between 17-46 amino acids from the N-terminal region of human TOB2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit