Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAIM3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520285

Catalog No. PIPA520285


Add to cart

Description

Description

Despite its predicted molecular weight, Toso often migrates at 60kDa in SDS-PAGE. A suggested positive control is human lung tissue lysate. PA5-20285 can be used with blocking peptide PEP-0403.

Apoptosis is an important process by which normal tissue homeostasis and function are maintained. One of the major signals that regulate this process is mediated by the activation of the Fas receptor by its ligand. This leads to the formation of a Fas-associated death domain (FADD)- containing death-inducing signaling complex and the activation of caspase-8, which in turn activates downstream effector caspases, such as caspase-3 and -7. Recent experiments have shown that overexpression of Toso, a novel regulator of Fas-induced apoptosis in lymphoid cells, in Jurkat cells as well as transgenic mice render these cells resistant to Fas-induced apoptosis but not to TRAIL-induced apoptosis. Furthermore, Toso was found to associate with FADD, suggesting that Toso functions by disrupting the formation of the death-inducing signaling complex.
Specifications

Specifications

FAIM3
Polyclonal
PBS with 0.02% sodium azide
O60667
FCMR, TOSO; Fc mu receptor; IgM Fc receptor; fas apoptotic inhibitory molecule 3; immunoglobulin mu Fc receptor; regulator of Fas-induced apoptosis Toso
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
9214
Western Blot
Unconjugated
FCMR
Liquid
FCMR
A 13 amino acid peptide from near the carboxy terminus of human Toso.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit