Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toxoplasma gondii Rabbit anti-Protozoa, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA17252

Catalog No. PA17252


Add to cart

Description

Description

PA1-7252 detects the Toxoplasma gondii. PA1-7252 has been successfully used in immunofluorescence procedures. The PA1-7252 immunogen is RH strain of Toxoplasma gondii. Store at 4°C for 6 months. For extended storage, store at -20°C.

Toxoplasma is a crescent shaped sporozoan that lives as intracellular parasite in various tissues of vertebrates and completes its life cycle in a single host. It is transmitted via raw/undercooked meat, contaminated soil, or by direct contact. Infection due to Toxoplasma gondii usually occurs in pregt women where a variable degree of immunosuppression may exist or in patients receiving immunosuppressive drug therapy following organ transplant. Toxoplasma infects tissue of the GI tract where an active infection is accompanied by fever and enlargement of the spleen. Symptoms of toxoplasmosis are generally mild but severe infection of lymph nodes may occur. Congenital toxoplasmosis, in which the maternal infection is transmitted during pregcy, can produce blindness or mental retardation in the newborn. Toxoplasma gondii infections in humans are widespread but vary greatly by region. The organism enters the intestinal epithelium and can spread to other host tissues. In healthy individuals the infection is usually asymptomatic. However, for individuals with compromised immune systems, a Toxoplasma infection can cause serious health problems.
Specifications

Specifications

Toxoplasma gondii
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Protozoa
Immunofluorescence
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
T. gondii
RH strain of Toxoplasma gondii.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit