Learn More
Toxoplasma gondii Goat anti-Protozoa, FITC, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Supplier:  Thermo Scientific PA173000
Catalog No. PIPA173000
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
The PA1-73000 antibody reacts with Toxoplasma gondii. PA1-73000 has been successfully used in Immunofluorescence applications. The PA1-73000 antibody was raised against the RH Strain of T. Gondii.
Toxoplasma is a crescent shaped sporozoan that lives as intracellular parasite in various tissues of vertebrates and completes its life cycle in a single host. It is transmitted via raw/undercooked meat, contaminated soil, or by direct contact. Infection due to Toxoplasma gondii usually occurs in pregt women where a variable degree of immunosuppression may exist or in patients receiving immunosuppressive drug therapy following organ transplant. Toxoplasma infects tissue of the GI tract where an active infection is accompanied by fever and enlargement of the spleen. Symptoms of toxoplasmosis are generally mild but severe infection of lymph nodes may occur. Congenital toxoplasmosis, in which the maternal infection is transmitted during pregcy, can produce blindness or mental retardation in the newborn. Toxoplasma gondii infections in humans are widespread but vary greatly by region. The organism enters the intestinal epithelium and can spread to other host tissues. In healthy individuals the infection is usually asymptomatic. However, for individuals with compromised immune systems, a Toxoplasma infection can cause serious health problems.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Toxoplasma gondii
Polyclonal
FITC
Goat
Purified
RUO
Protozoa
Antibody
IgG
Immunofluorescence
4-5 mg/mL
T. gondii
RH Strain of T. Gondii.
1 mL
Primary
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Liquid
Recommendations
Recommendations