Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toxoplasma gondii MIC3 Mouse anti-Protozoa, Clone: A80D, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA518267

Catalog No. PIMA518267


Add to cart

Description

Description

This antibody recognizes the MIC 3 protein of T. gondii. Purity is >90%.

Toxoplasma is a crescent shaped sporozoan that lives as intracellular parasite in various tissues of vertebrates and completes its life cycle in a single host. It is transmitted via raw/undercooked meat, contaminated soil, or by direct contact. Infection due to Toxoplasma gondii usually occurs in pregt women where a variable degree of immunosuppression may exist or in patients receiving immunosuppressive drug therapy following organ transplant. Toxoplasma infects tissue of the GI tract where an active infection is accompanied by fever and enlargement of the spleen. Symptoms of toxoplasmosis are generally mild but severe infection of lymph nodes may occur. Congenital toxoplasmosis, in which the maternal infection is transmitted during pregcy, can produce blindness or mental retardation in the newborn. Toxoplasma gondii infections in humans are widespread but vary greatly by region. The organism enters the intestinal epithelium and can spread to other host tissues. In healthy individuals the infection is usually asymptomatic. However, for individuals with compromised immune systems, a Toxoplasma infection can cause serious health problems.
Specifications

Specifications

Toxoplasma gondii MIC3
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
T. gondii; T. gondii MIC3
Toxoplasma gondii native protein
Protein A
RUO
Antibody
Monoclonal
ELISA, Immunofluorescence, Western Blot
A80D
Unconjugated
Liquid
Mouse
IgG1
100 μg
4° C
Primary
Protozoa
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit