Learn More
C20orf10 Mouse anti-Human, Clone: 3C2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA521720
Catalog No. PIMA521720
Add to cart
Description
Description
Peptide Sequence: RNRTPMGHMK QLDVADQWIW FEGLPTRIHL PAPRVMCRSS TLRWVKRRCT RFCSASLEMP MWHPYKVDVT WTRARGASRG WRSRHQLKGR NGWRNSRVYK
May play a significant role in p53/TP53-mediating signaling pathway.
Specifications
Specifications
C20orf10
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9Y2B4
P3H3
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
27296
ELISA
3C2
Unconjugated
TP53TG5
Liquid
TP53TG5
TP53TG5 (AAH36785, 191 a.a. ∽ 290 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human