Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAPPC6A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525211

Catalog No. PIPA525211


Add to cart

Description

Description

TRAPPC6A is a subunit of the TRAPP I and II complexes.
Specifications

Specifications

TRAPPC6A
Polyclonal
PBS with 0.09% sodium azide
O75865
TRS33, TRAPP complex subunit 6A, TRAPPC6Adelta29-42, trafficking protein particle complex subunit 6A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79090
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
TRAPPC6A
Liquid
TRAPPC6A
KLH conjugated synthetic peptide between 34-62 amino acids from the N-terminal region of human TPC6A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit