Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TPGS2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524031
Description
The function of C18orf10 remains unknown.Specifications
TPGS2 | |
Polyclonal | |
TPGS2 | |
C18orf10, HsT3006, L17, PGs2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25941 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q68CL5 | |
TPGS2 | |
KLH conjugated synthetic peptide between 109-137 amino acids from the central region of human TPGS2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction