Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPK1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526150

Catalog No. PIPA526150


Add to cart

Description

Description

TPK1 is a protein, that exists as a homodimer, which catalyzes the conversion of thiamine to thiamine pyrophosphate.
Specifications

Specifications

TPK1
Polyclonal
PBS with 0.09% sodium azide
Q9H3S4, Q9R0M5
HTPK1, PP20, placental protein 20, thiamine diphosphokinase, thiamine kinase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27010, 29807
Immunohistochemistry, Western Blot
Unconjugated
TPK1
Liquid
TPK1
KLH conjugated synthetic peptide between 34-61 amino acids from the N-terminal region of human TPK1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit