Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tropomyosin 3, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length recombinant TPM3.

Manufacturer:  Abnova Corporation H00007170A01

Catalog No. 89-004-349


Add to cart

Description

Description

This gene encodes a member of the tropomyosin family of actin-binding proteins involved in the contractile system of striated and smooth muscles and the cytoskeleton of nonmuscle cells. Tropomyosins are dimers of coiled-coil proteins that polymerize end-to-end along the major groove in most actin filaments. They provide stability to the filaments and regulate access of other actin-binding proteins. In muscle cells, they regulate muscle contraction by controlling the binding of myosin heads to the actin filament. Mutations in this gene result in autosomal dominant nemaline myopathy, and oncogenes formed by chromosomal translocations involving this locus are associated with cancer. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MAGITTIEAVKRKIQVLQQQADDAEERAERLQREVEGERRAREQAEAEVASLNRRIQLVEEELDRAQERLATALQKLEEAEKAADESERGMKVIENRALKDEEKMELQEIQLKEAKHIAEEADRKYEEVARKLVIIEGDLERTEERAELAESRCREMDEQIRLMDQNLKRLSAAEEKYSQKEDKYEGEIKILTDKLKEAETRAEFAERSVAKLEKTIDDLEDKLKCTKEEHLCTQRMLDQTLLDLNEM
Specifications

Specifications

tropomyosin 3
Unconjugated
50% glycerol
BC015403
FLJ41118|MGC14582|MGC3261|MGC72094|NEM1|OK/SW-cl.5|TM-5|TM3|TM30|TM30nm|TPMsk3|TRK|hscp30
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
7170
ELISA, Western Blot
Mouse polyclonal antibody raised against a full-length recombinant TPM3.
TPM3
Serum
TPM3
TPM3 (AAH15403, 1 a.a. ~ 248 a.a) full-length recombinant protein with GST tag.
RUO
Y
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit