Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPRG1L Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525005

Catalog No. PIPA525005

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TPRG1L
Polyclonal
PBS with 0.09% sodium azide
Q5T0D9, Q9DBS2
FAM79A, RP11-46F15.3, h-mover, mover, family with sequence similarity 79, member A, mossy fiber terminal-associated vertebrate-specific presynaptic protein, tumor protein p63-regulated gene 1-like protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
127262, 67808
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TPRG1L
Liquid
TPRG1L, Tprgl
KLH conjugated synthetic peptide between 35-63 amino acids from the N-terminal region of human TPRGL
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit