Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THRA, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521337

Catalog No. PIPA521337

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA5-21337 targets TR Alpha in WB applications and shows reactivity with Human samples.The PA5-21337 immunogen is a recombinant fragment corresponding to a region within amino acids 317 and 490 of Thyroid Hormone Receptor alpha 2 (Uniprot ID#P10827).
Specifications

Specifications

TR Alpha
Polyclonal
Unconjugated
THRA
Purified
THRA
Recombinant fragment corresponding to a region within amino acids 317 and 490 of Thyroid Hormone Receptor alpha 2 (Uniprot ID#P10827)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.33mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
P10827
AR7, CHNG6, EAR7, ERB-T-1, ERBA, ERBA1, NR1A1, THRA1, THRA2, c-ERBA-1, EAR-7, ERBA-related 7, V-erbA-related protein 7, c-erbA-alpha, nuclear receptor subfamily 1 group A member 1, thyroid hormone receptor alpha, thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a) oncogene homolog, avian), thyroid normone nuclear receptor alpha variant 1, triiodothyronine receptor
Rabbit
IgG
100μL
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
7067
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit