Learn More
anti-TR2/NR2C1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP130047
Catalog No. NBP130047
Add to cart
Description
Description
TR2/NR2C1 Polyclonal antibody specifically detects TR2/NR2C1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
TR2/NR2C1
Unconjugated
Neat whole antisera with No Preservative
Antisera
NR2C1
Fusion protein from the N-terminal region of mouse TR2
64 kDa
0.1 ml
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Specific for the ~64 kDa TR2 protein in Western blots of testes and nuclear extracts from MEL cell lines.
Western Blot
Western Blot 1:1000
Q505F1
nuclear receptor subfamily 2 group C member 1, nuclear receptor subfamily 2, group C isoform, nuclear receptor subfamily 2, group C, member 1, Orphan nuclear receptor TR2, Testicular receptor 2, TR2, TR2 nuclear hormone receptor, TR2-11
Rabbit
IgG
Unpurified
RUO
Primary
7181
Human, Mouse, Rat