Learn More
TRA-1-85 Mouse anti-Human, Brilliant Violet 421, Clone: TRA-1-85, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 563302
Catalog No. BDB563302
Add to cart
Description
Description
The TRA-1-85 monoclonal antibody (mAb) specifically binds to the high frequency blood group antigen OKa. The OKa blood group antigen is a 35-69 kDa glycoprotein also known as CD147, EMMPRIN, M6, or Basigin. The OKa group is widely expressed on most human tissues including, but not limited to kidney, peripheral blood leukocytes, liver, pancreas, colon, skin, and brain. TRA-1-85 mAb is often used to discern human cells from non-human cells in multiple applications including animal transplantation studies. During development, the purified TRA-1-85 mAb was found to detect OKa blood group antigen by western blot analysis of cellular lysates, by immunofluorescent staining of fixed cells, and by flow cytometric analysis of live cells.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.
Specifications
Specifications
TRA-1-85
TRA-1-85
OKa, CD147, EMMPRIN, Basigin
Affinity Purified
Human Embryonal Carcinoma Cell Line
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human