Learn More
TAAR3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00081W
Catalog No. PIOSR00081W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material. OSR00081W can be used with blocking peptide GST00003P.
Not expressed in the pons, thalamus, globus pallidus, caudate, putamen or cerebellum. TAAR3 might be the product of a pseudogene.
Specifications
Specifications
TAAR3
Polyclonal
whole serum with no preservative
Q9P1P4
Putative trace amine-associated receptor 3, G-protein coupled receptor 57, TAAR3, GPR57. CAUTION: Could be the product of a pseudogene.
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
TAAR3P
Lyophilized
TAAR3P
A synthetic peptide from c-terminal region of human trace amine-associated receptor 3
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
9288