Learn More
TAAR6 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00082W
Catalog No. PIOSR00082W
Add to cart
Description
Description
This antibody is predicted to react with human based on sequence homology. OSR00082W can be used with blocking peptide GST00005P.
Trace amines are biogenic amines present in very low levels in mammalian tissues. Although some trace amines have clearly defined roles as neurotransmitters in invertebrates, the extent to which they function as true neurotransmitters in vertebrates has remained speculative. Trace amines are likely to be involved in a variety of physiological functions that have yet to be fully understood. Expressed at low abundance in various brain tissues, as well as in fetal liver, but not in the cerebellum or placenta. In the brain, comparable levels of expression in basal ganglia, frontal cortex, substantia nigra, amygdala and hippocampus, highest expression in hippocampus and lowest expression in basal ganglia.
Specifications
Specifications
TAAR6
Polyclonal
whole serum with no preservative
Q5QD13, Q923Y5
Trace amine-associated receptor 6, Trace amine receptor 4, TaR-4, TA4, TAR4, TRAR4, Ta4, Tar4, Trar4
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
Taar6
Lyophilized
TAAR6
A synthetic peptide from mouse TAAR6 conjugated to an immunogenic carrier protein
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
215855, 294124