Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRADD Mouse, Unlabeled, Clone: 37, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610573

 View more versions of this product

Catalog No. BDB610573


Add to cart

Description

Description

TNFα (Tumor Necrosis Factor) stimulates programmed cell death and NF-κB activation as a result of its binding to the TNF receptor 1 (TNFR1). Within this receptor, a sequence referred to as the “death domain” has been shown to be necessary for both of these functions. Using the yeast two-hybrid system to detect proteins which interact with the receptor through this “death domain”, a 34kDa protein was found and designated TRADD (TNFR1-Associated Death Domain protein). TRADD appears to contain no intrinsic catalytic activity. It also contains a death domain and it has been shown to bind to FADD and RIP. Mutational analysis of TRADD demonstrates that programmed cell death and NF-κB activation are distinct and controlled independently.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

TRADD
37
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
TNFR1-Associated Death Domain protein
Affinity Purified
Aqueous buffered solution containing BSA
150μg
Cell Biology
Primary
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.