Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRADD Mouse, Unlabeled, Clone: 37, BD

Mouse Monoclonal Antibody

$211.00

Specifications

Antigen TRADD
Isotype IgG1
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB610572
View Documents Promotion Details
BD Biosciences
610572
50μg
Each for $211.00
Add to cart
 
BDB610573
View Documents Promotion Details
BD Biosciences
610573
150μg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

TNFα (Tumor Necrosis Factor) stimulates programmed cell death and NF-κB activation as a result of its binding to the TNF receptor 1 (TNFR1). Within this receptor, a sequence referred to as the “death domain” has been shown to be necessary for both of these functions. Using the yeast two-hybrid system to detect proteins which interact with the receptor through this “death domain”, a 34kDa protein was found and designated TRADD (TNFR1-Associated Death Domain protein). TRADD appears to contain no intrinsic catalytic activity. It also contains a death domain and it has been shown to bind to FADD and RIP. Mutational analysis of TRADD demonstrates that programmed cell death and NF-κB activation are distinct and controlled independently.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

TRADD
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Unconjugated
Cell Biology
Mouse
Affinity Purified
Monoclonal
TNFR1-Associated Death Domain protein
IgG1
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Western Blot
RUO
37
250μg/mL
Store undiluted at -20°C.
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit