Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TRAF-2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication

Manufacturer:  Novus Biologicals NB10056172

 View more versions of this product

Catalog No. NB10056172


Add to cart

Description

Description

TRAF-2 Polyclonal specifically detects TRAF-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

TRAF-2
Unconjugated
Whole antisera with 0.05% Sodium Azide
E3 ubiquitin-protein ligase TRAF2, EC 6.3.2.-, MGC:45012, TNF receptor-associated factor 2, TRAP3TRAP, tumor necrosis factor type 2 receptor associated protein 3, Tumor necrosis factor type 2 receptor-associated protein 3
Rabbit
IgG
0.05 ml
Apoptosis, Cancer, Cytokine Research, Zinc Finger
Primary
7186
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Western Blot 1/1000 - 1/200, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin 1/1000 - 1/500, Immunohistochemistry-Frozen Use at an assay dependent dilution
Antisera
TRAF2
A synthetic peptide corresponding to amino acids 87-106 (SSSAFPDNAARREVESLPAV) of human TRAF2 was used as immunogen, GenBank no. NP_066961.2. This peptide sequence is 100% conserved in mouse and rat TRAF2. The peptide immunogen sequence is 100% conserved in multiple human TRAF2 GenBank accession numbers. These TRAF2 accession numbers are of varying amino acid (aa) lengths and include: 1. NP_066961.2 = 501 aa. 2. CAI12705.1 =121 aa. 3. CAI12704.1 = 422 aa. 4. CAI15104.1 = 193 aa. 5. BAD96681.1 = 205 aa.
Unpurified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
The gene encoding TRAF2 produces, by alternative splicing, 15 different transcripts, all with introns, putatively encoding 15 different protein isoforms. These isoforms are of varying amino acid lengths. Therefore the molecular weight observed by western blot may vary, depending on the isoform expressed. Users are encouraged to use the peptide immunogen sequence information provided in the antigen section to gain additional information about the potential TRAF2 isoforms detected by the antibody.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only