Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TRAF-4, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP147306

Catalog No. NBP147306

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TRAF-4
1.0 mg/mL
Immunoprecipitation 2-5 ug/mg, Immunohistochemistry-Paraffin 1:500-1:2000
Affinity Purified
TRAF4
The immunogen for this product maps to a region between residue 420 and 470 of human TNF Receptor-Associated Factor 4 using the numbering given in entry NP_004286.2 (GeneID 9618).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Human
Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
CART1RING finger protein 83, Cysteine-rich domain associated with RING and Traf domains protein 1, Metastatic lymph node gene 62 protein, MLN 62, MLN62cysteine-rich domain associated with ring and TRAF domain, RNF83malignant 62, TNF receptor-associated factor 4, tumor necrosis receptor-associated factor 4A
Rabbit
IgG
100 ul
Cancer, Zinc Finger
Primary
9618
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only