TNF receptor-associated factor 4, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human TRAF4 protein.

Manufacturer: Abnova Corporation H00009618D01P

Catalog No. 89-018-179

  Check Availability
Add to Cart

    Specifications

    Formulation In 1X PBS, pH 7.4
    Gene TRAF4
    Gene Accession No. NM_004295
    Gene Alias CART1, MLN62, RNF83
    Gene Symbols TRAF4
    Applications In situ PLA,Western Blotting
    Storage Requirements Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
    Gene ID (Entrez) 9618
    Species Reactivity Human
    Description TNF receptor-associated factor 4
    Antigen TNF receptor-associated factor 4
    Format Affinity Purified
    Host Species Rabbit
    Immunogen TRAF4 (NP_004286.2, 1 a.a. ∽470 a.a) full-length human protein.
    Purification Method Protein A column
    Quantity 100μg
    Regulatory Status RUO
    Primary or Secondary Primary
    Monoclonal or Polyclonal Polyclonal
    Conjugate Unlabeled
    Cross Reactivity Human

    This gene encodes a member of the TNF receptor associated factor (TRAF) family. TRAF proteins are associated with, and mediate signal transduction from members of the TNF receptor superfamily. The encoded protein has been shown to interact with neurotrophin receptor, p75 (NTR/NTSR1), and negatively regulate NTR induced cell death and NF-kappa B activation. This protein has been found to bind to p47phox, a cytosolic regulatory factor included in a multi-protein complex known as NAD(P)H oxidase. This protein thus, is thought to be involved in the oxidative activation of MAPK8/JNK. Alternatively spliced transcript variants have been observed but the full-length nature of only one has been determined. [provided by RefSeq

    Sequence: MPGFDYKFLEKPKRRLLCPLCGKPMREPVQVSTCGHRFCDTCLQEFLSEGVFKCPEDQLPLDYAKIYPDPELEVQVLGLPIRCIHSEEGCRWSGPLRHLQGHLNTCSFNVIPCPNRCPMKLSRRDLPAHLQHDCPKRRLKCEFCGCDFSGEAYESHEGMCPQESVYCENKCGARMMRRLLAQHATSECPKRTQPCTYCTKEFVFDTIQSHQYQCPRLPVACPNQCGVGTVAREDLPGHLKDSCNTALVLCPFKDSGCKHRCPKLAMARHVEESVKPHLAMMCALVSRQRQELQELRRELEELSVGSDGVLIWKIGSYGRRLQEAKAKPNLECFSPAFYTHKYGYKLQVSAFLNGNGSGEGTHLSLYIRVLPGAFDNLLEWPFARRVTFSLLDQSDPGLAKPQHVTETFHPDPNWKNFQKPGTWRGSLDESSLGFGYPKFISHQDIRKRNYVRDDAVFIRAAVELPRKILS

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit