Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TNF receptor-associated factor 5, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human TRAF5 protein.

Manufacturer:  Abnova Corporation H00007188B01

Catalog No. 89-006-394


Add to cart

Description

Description

The scaffold protein encoded by this gene is a member of the tumor necrosis factor receptor-associated factor (TRAF) protein family and contains a meprin and TRAF homology (MATH) domain, a RING-type zinc finger, and two TRAF-type zinc fingers. TRAF proteins are associated with, and mediate signal transduction from members of the TNF receptor superfamily. This protein is one of the components of a multiple protein complex which binds to tumor necrosis factor (TNF) receptor cytoplasmic domains and mediates TNF-induced activation. Alternate transcriptional splice variants have been characterized. [provided by RefSeq

Sequence: MAYSEEHKGMPCGFIRQNSGNSISLDFEPSIEYQFVERLEERYKCAFCHSVLHNPHQTGCGHRFCQHCILSLRELNTVPICPVDKEVIKSQEVFKDNCCKREVLNLYVYCSNAPGCNAKVILGRYQDHLQQCLFQPVQCSNEKCREPVLRKDLKEHLSASCQFRKEKCLYCKKDVVVINLQNHEENLCPEYPVFCPNNCAKIILKTEVDEHLAVCPEAEQDCPFKHYGCAVTDKRRNLQQHEHSALREHMRLVLEKNVQLEEQISDLHKSLEQKESKIQQLAETIKKLEKEFKQFAQLFGKNGSFLPNIQVFASHIDKSAWLEAQVHQLLQMVNQQQNKFDLRPLMEAVDTVKQKITLLENNDQRLAVLEEETNKHDTHINIHKAQLSKNEERFKLLEGTCYNGKLIWKVTDYKMKKREAVDGHTVSIFSQSFYTSRCGYRLCARAYLNGDGSGRGSHLSLYFVVMRGEFDSLLQWPFRQRVTLMLLDQSGKKNIMETFKPDPNSSSFKRPDGEMNIASGCPRFVAHSVLENAKNAYIKDDTLFLKVAVDLTDLEDL
Specifications

Specifications

TNF receptor-associated factor 5
Unconjugated
No additive
NM_001033910
MGC:39780|RNF84
Mouse
50 uL
Apoptosis
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human TRAF5 protein.
TRAF5
Serum
TRAF5
TRAF5 (NP_001029082, 1 a.a. ~ 557 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
7188
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit