Learn More
TRAF6 Rabbit anti-Human, Horse, Rhesus Macaque, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061110
Catalog No. 06-111-0MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect TRAF6 using this Anti-TRAF6 Antibody validated for use in western blot, immunohistochemistry.
Specifications
Specifications
TRAF6
Unconjugated
Purified
Rabbit
Protein A purfied
2-8°C for 12 months
Polyclonal
Equine, Human, Rhesus
Immunohistochemistry, Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
TRAF6; RNF85
KLH-conjugated linear peptide corresponding to the MATH (meprin-associated Traf homology) domain of human TRAF6.
100μg
Primary
NP_004611