Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAIL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14172

Catalog No. PA14172


Add to cart

Description

Description

PA1-4172 detects TRAIL in human samples. PA1-4172 has been successfully used in Western blot procedures. The PA1-4172 immunogen is a synthetic peptide corresponding to the C-terminal residues M(223) K S A R N S C W S K D A(235) of human TRAIL.

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including tumor necrosis factor (TNF) and Fas ligand in the TNF family through their death domain containing receptors, TNF receptor 1 (TNFR1) and Fas. Another member in the TNF family has been identified and designated TRAIL (for TNF-related apoptosis inducing ligand) and Apo-2L (for Apo-2 ligand). Receptors for TRAIL include two death domain containing receptors, death receptor 4 (DR4) and DR5, as well as two decoy receptors, DcR1 and DcR2, lacking the intracellular signaling death domain. TRAIL is a type II membrane protein and expressed in a variety of human tissues. Like TNF and Fas ligand, TRAIL induces apoptosis and NF-kB activation in many tissues and cells.
Specifications

Specifications

TRAIL
Polyclonal
Unconjugated
TNFSF10
Liquid
TNFSF10
Synthetic peptide corresponding to the C-terminal residues M(223) K S A R N S C W S K D A(235) of human TRAIL.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P50591
Apo2L, Apo-2 Ligand, TNF-Related Apoptosis-Inducing Ligand, Tumor necrosis factor ligand, superfamily member 10, TRAIL protein, CD253 antigen, TNFSF10
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
8743
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit