Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRAIL R3/TNFRSF10C Mouse anti-Human, Clone: 90905, R&D Systems™

Mouse Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen TRAILR3/TNFRSF10C
Immunogen Mouse myeloma cell line NS0-derived recombinant human TRAIL R3/TNFRSF10C Ala26-Ala221 Accession # O14798
Clone 90905
Applications Western Blot, Immunohistochemistry, ELISA
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB6301SP
View Documents
R&D Systems
MAB6301SP
25 ug
Each for $114.50
Add to cart
 
MAB6301100
View Documents
R&D Systems
MAB6301100
100 ug
Each for $377.00
Add to cart
 
MAB6301500
View Documents
R&D Systems
MAB6301500
500 ug
Each for $839.00
Add to cart
 
Description

Description

TRAIL R3/TNFRSF10C Monoclonal antibody MAB6301 Specifically detects TRAIL R3/TNFRSF10C in Human samples. It is validated for ELISA Capture, Immunohistochemistry, Western Blotting
Specifications

Specifications

TRAILR3/TNFRSF10C
90905
Unconjugated
O14798
Monoclonal
Primary
Detects human TRAIL R3/TNFRSF10C in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) TRAIL R1, rhTRAIL R2, or rhTRAIL R4 is observed.
Protein A or G purified from ascites
Mouse myeloma cell line NS0-derived recombinant human TRAIL R3/TNFRSF10C Ala26-Ala221 Accession # O14798
Western Blot, Immunohistochemistry, ELISA
Purified
Mouse
IgG1
8794
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit